Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTCACTAAGATGGAGTTTTAG[T/A]GGAAAAGCAGAGCAATTATATTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616347 MIM: 607716 | ||||||||||||||||||||
Literature Links: |
PRDM11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRDM11 - PR domain 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT13 - synaptotagmin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247987.1 | 4632 | UTR 3 | NP_001234916.1 | |||
NM_020826.2 | 4632 | UTR 3 | NP_065877.1 |