Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGACTATGAAAATGACTGTCTC[A/G]TTCACCTCACGGCGACCCCGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRAS - HRas proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101059906 - collagen alpha-2(IV) chain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_003959933.4 | Intron | XP_003959982.2 |
LRRC56 - leucine rich repeat containing 56 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198075.3 | Intron | NP_932341.1 | ||||
XM_005252775.2 | Intron | XP_005252832.1 | ||||
XM_005252776.2 | Intron | XP_005252833.1 | ||||
XM_006718132.2 | Intron | XP_006718195.1 | ||||
XM_006718133.2 | Intron | XP_006718196.1 | ||||
XM_011519875.2 | Intron | XP_011518177.1 | ||||
XM_011519876.1 | Intron | XP_011518178.1 | ||||
XM_011519877.2 | Intron | XP_011518179.1 | ||||
XM_017017167.1 | Intron | XP_016872656.1 | ||||
XM_017017168.1 | Intron | XP_016872657.1 |