Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACATCTCAGAGGTCCAAGTGGT[G/T]AGCTGTTGTCCAGCCTCCCGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613528 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR302E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR302E - microRNA 302e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT9 - synaptotagmin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175733.3 | Intron | NP_783860.1 | ||||
XM_011519900.2 | Intron | XP_011518202.1 | ||||
XM_011519901.2 | Intron | XP_011518203.1 | ||||
XM_011519902.2 | Intron | XP_011518204.1 | ||||
XM_011519904.2 | Intron | XP_011518206.1 | ||||
XM_011519906.2 | Intron | XP_011518208.1 | ||||
XM_011519907.2 | Intron | XP_011518209.1 |