Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGTCAATTACTTCACCAATATTT[A/G]CAGCAGTTGCTTCAACTCTTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYB5R2 - cytochrome b5 reductase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OVCH2 - ovochymase 2 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198185.4 | Intron | NP_937828.3 | ||||
XM_017017659.1 | Intron | XP_016873148.1 |