Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCCATTCTATTCTCAGGCTACA[C/T]CCTGGTCTCAGTTATCAACGCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASC23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASC23 - cancer susceptibility candidate 23 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUB - tubby bipartite transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003320.4 | Intron | NP_003311.2 | ||||
NM_177972.2 | Intron | NP_813977.1 | ||||
XM_005253109.3 | Intron | XP_005253166.1 | ||||
XM_011520344.2 | Intron | XP_011518646.1 |