Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAAAAGTTAAGGCACCCTCTG[C/T]CACTCCCCACTCCCACTGAATCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST8SIA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST8SIA1 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304450.1 | 8542 | UTR 3 | NP_001291379.1 | |||
NM_003034.3 | 8542 | UTR 3 | NP_003025.1 |