Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGTCTTGAGCGGCTGCTCTTT[G/T]TACGTCTCCACCTTGTTAGTTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 168470 | ||||||||||||||||||||
Literature Links: |
PTHLH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTHLH - parathyroid hormone like hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002820.2 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | NP_002811.1 | |
NM_198964.1 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | NP_945315.1 | |
NM_198965.1 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | NP_945316.1 | |
NM_198966.1 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | NP_945317.1 | |
XM_011520774.2 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | XP_011519076.1 | |
XM_011520775.2 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | XP_011519077.1 | |
XM_017019674.1 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | XP_016875163.1 | |
XM_017019675.1 | 645 | Nonsense Mutation | TAA,TAC | *,Y 114 | XP_016875164.1 |