Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCACTCAGCCCTATGGATTTC[A/G]CTTACAGGAAACGGTAGAATGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600618 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETV6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETV6 - ETS variant 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001987.4 | Intron | NP_001978.1 | ||||
XM_011520607.2 | Intron | XP_011518909.1 | ||||
XM_011520608.2 | Intron | XP_011518910.1 | ||||
XM_011520609.2 | Intron | XP_011518911.1 | ||||
XM_011520611.2 | Intron | XP_011518913.1 | ||||
XM_011520612.2 | Intron | XP_011518914.1 | ||||
XM_017018990.1 | Intron | XP_016874479.1 | ||||
XM_017018991.1 | Intron | XP_016874480.1 |