Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCTAAAACAAAGGCCGATGAA[A/G]TAATCAGTTCCCAGACTGGGTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613541 MIM: 605501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf65 - chromosome 12 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143905.2 | Intron | NP_001137377.1 | ||||
NM_001194995.1 | Intron | NP_001181924.1 | ||||
NM_152269.4 | Intron | NP_689482.1 | ||||
XM_005253630.4 | Intron | XP_005253687.1 | ||||
XM_011538980.2 | Intron | XP_011537282.1 |
MPHOSPH9 - M-phase phosphoprotein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022782.3 | Intron | NP_073619.3 | ||||
XM_011537740.2 | Intron | XP_011536042.1 | ||||
XM_011537741.2 | Intron | XP_011536043.1 | ||||
XM_017018673.1 | Intron | XP_016874162.1 | ||||
XM_017018674.1 | Intron | XP_016874163.1 | ||||
XM_017018675.1 | Intron | XP_016874164.1 | ||||
XM_017018676.1 | Intron | XP_016874165.1 | ||||
XM_017018677.1 | Intron | XP_016874166.1 | ||||
XM_017018678.1 | Intron | XP_016874167.1 | ||||
XM_017018679.1 | Intron | XP_016874168.1 | ||||
XM_017018680.1 | Intron | XP_016874169.1 | ||||
XM_017018681.1 | Intron | XP_016874170.1 |