Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAGCCGAAGCAGACATTCCAAG[G/T]GGGCAGCTCAAACAAAACCCAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RILPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RILPL1 - Rab interacting lysosomal protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319243.1 | Intron | NP_001306172.1 | ||||
NM_001319244.1 | Intron | NP_001306173.1 | ||||
NM_001319302.1 | Intron | NP_001306231.1 | ||||
NM_178314.4 | Intron | NP_847884.2 | ||||
XM_006719372.2 | Intron | XP_006719435.1 | ||||
XM_006719373.3 | Intron | XP_006719436.1 | ||||
XM_011538282.2 | Intron | XP_011536584.1 | ||||
XM_011538284.2 | Intron | XP_011536586.1 | ||||
XM_011538285.2 | Intron | XP_011536587.1 |
SNRNP35 - small nuclear ribonucleoprotein U11/U12 subunit 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |