Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATTTTATGAATTGTTTCCAAGT[C/T]TGGGCATTTGTTTGTTTGATCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609509 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL31 - interleukin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014336.1 | Intron | NP_001014358.1 |
LRRC43 - leucine rich repeat containing 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098519.1 | Intron | NP_001091989.1 | ||||
NM_152759.4 | Intron | NP_689972.3 | ||||
XM_017019124.1 | Intron | XP_016874613.1 | ||||
XM_017019125.1 | Intron | XP_016874614.1 |