Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATTCCTGAAAATCATGGTGAG[A/G]ACATGAGGAAAATGTCCTTCCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608109 MIM: 603168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PUS1 - pseudouridylate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002019.2 | 852 | Missense Mutation | AAC,GAC | N,D 105 | NP_001002019.1 | |
NM_001002020.2 | 852 | Missense Mutation | AAC,GAC | N,D 105 | NP_001002020.1 | |
NM_025215.5 | 852 | Missense Mutation | AAC,GAC | N,D 133 | NP_079491.2 | |
XM_011538768.2 | 852 | UTR 5 | XP_011537070.1 | |||
XM_011538769.2 | 852 | Missense Mutation | AAC,GAC | N,D 133 | XP_011537071.1 |
ULK1 - unc-51 like autophagy activating kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |