Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCAAGAACCTCAGTCAATGAAA[A/G]AGAAATGGTGGAATGTTTGACAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604689 MIM: 603834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP3 - A-kinase anchoring protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA9 - NADH:ubiquinone oxidoreductase subunit A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005002.4 | Intron | NP_004993.1 |