Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGTGGCTCCGGCTCCTCCTCT[C/G]TCACCCCCAATAGCGGTGGGACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611055 | ||||||||||||||||||||
Literature Links: |
LINC01089 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01089 - long intergenic non-protein coding RNA 1089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1B - SET domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015048.1 | 892 | Missense Mutation | CTC,GTC | L,V 243 | NP_055863.1 | |
XM_005253858.4 | 892 | Missense Mutation | CTC,GTC | L,V 243 | XP_005253915.1 | |
XM_006719296.3 | 892 | Missense Mutation | CTC,GTC | L,V 243 | XP_006719359.1 |