Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGCTCTATCACGAATTAGTGA[C/T]GAGTTCTGGCTAGACCCATCTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608368 MIM: 606932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD9B - RAD9 checkpoint clamp component B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286531.1 | 125 | UTR 5 | NP_001273460.1 | |||
NM_001286532.1 | 125 | UTR 5 | NP_001273461.1 | |||
NM_001286533.1 | 125 | UTR 5 | NP_001273462.1 | |||
NM_001286534.1 | 125 | UTR 5 | NP_001273463.1 | |||
NM_001286535.1 | 125 | Silent Mutation | GAC,GAT | D,D 29 | NP_001273464.1 | |
NM_001286536.1 | 125 | Silent Mutation | GAC,GAT | D,D 29 | NP_001273465.1 | |
NM_152442.3 | 125 | Silent Mutation | GAC,GAT | D,D 29 | NP_689655.3 | |
XM_005253849.4 | 125 | Missense Mutation | GAC,GAT | D,D 29 | XP_005253906.1 | |
XM_011537971.2 | 125 | Missense Mutation | GAC,GAT | D,D 29 | XP_011536273.1 | |
XM_011537972.2 | 125 | Missense Mutation | GAC,GAT | D,D 29 | XP_011536274.1 | |
XM_011537974.2 | 125 | Intron | XP_011536276.1 | |||
XM_017018877.1 | 125 | Missense Mutation | GAC,GAT | D,D 29 | XP_016874366.1 | |
XM_017018878.1 | 125 | UTR 5 | XP_016874367.1 | |||
XM_017018879.1 | 125 | UTR 5 | XP_016874368.1 | |||
XM_017018880.1 | 125 | UTR 5 | XP_016874369.1 | |||
XM_017018881.1 | 125 | UTR 5 | XP_016874370.1 | |||
XM_017018882.1 | 125 | UTR 5 | XP_016874371.1 |
VPS29 - VPS29, retromer complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |