Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCCCCTCACACACAAACACAC[A/G]TGTCCTGTTGAACTCATGCACGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602582 MIM: 604118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTX1 - deltex E3 ubiquitin ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004416.2 | 3117 | UTR 3 | NP_004407.2 | |||
XM_011538009.2 | 3117 | UTR 3 | XP_011536311.1 |
RASAL1 - RAS protein activator like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193520.1 | 3117 | Intron | NP_001180449.1 | |||
NM_001193521.1 | 3117 | Intron | NP_001180450.1 | |||
NM_001301202.1 | 3117 | Intron | NP_001288131.1 | |||
NM_004658.2 | 3117 | Intron | NP_004649.2 | |||
XM_005253950.4 | 3117 | UTR 3 | XP_005254007.1 | |||
XM_006719641.3 | 3117 | UTR 3 | XP_006719704.1 | |||
XM_006719642.3 | 3117 | UTR 3 | XP_006719705.1 | |||
XM_011538852.2 | 3117 | UTR 3 | XP_011537154.1 | |||
XM_011538853.2 | 3117 | UTR 3 | XP_011537155.1 | |||
XM_011538854.2 | 3117 | UTR 3 | XP_011537156.1 | |||
XM_017020028.1 | 3117 | UTR 3 | XP_016875517.1 | |||
XM_017020029.1 | 3117 | UTR 3 | XP_016875518.1 | |||
XM_017020030.1 | 3117 | UTR 3 | XP_016875519.1 | |||
XM_017020031.1 | 3117 | UTR 3 | XP_016875520.1 |