Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACATATCATAGAGAGCTTCCCTG[G/T]ATTGTGTTTGAGCCATTTCCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601515 MIM: 604234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF14 - fibroblast growth factor 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321931.1 | 1835 | UTR 3 | NP_001308860.1 | |||
NM_001321932.1 | 1835 | UTR 3 | NP_001308861.1 | |||
NM_001321933.1 | 1835 | UTR 3 | NP_001308862.1 | |||
NM_001321934.1 | 1835 | UTR 3 | NP_001308863.1 | |||
NM_001321935.1 | 1835 | UTR 3 | NP_001308864.1 | |||
NM_001321936.1 | 1835 | UTR 3 | NP_001308865.1 | |||
NM_001321937.1 | 1835 | Intron | NP_001308866.1 | |||
NM_001321938.1 | 1835 | UTR 3 | NP_001308867.1 | |||
NM_001321939.1 | 1835 | UTR 3 | NP_001308868.1 | |||
NM_001321940.1 | 1835 | UTR 3 | NP_001308869.1 | |||
NM_001321941.1 | 1835 | UTR 3 | NP_001308870.1 | |||
NM_001321942.1 | 1835 | UTR 3 | NP_001308871.1 | |||
NM_001321943.1 | 1835 | UTR 3 | NP_001308872.1 | |||
NM_001321944.1 | 1835 | UTR 3 | NP_001308873.1 | |||
NM_001321945.1 | 1835 | UTR 3 | NP_001308874.1 | |||
NM_001321946.1 | 1835 | UTR 3 | NP_001308875.1 | |||
NM_001321947.1 | 1835 | UTR 3 | NP_001308876.1 | |||
NM_001321948.1 | 1835 | UTR 3 | NP_001308877.1 | |||
NM_001321949.1 | 1835 | UTR 3 | NP_001308878.1 | |||
NM_004115.3 | 1835 | UTR 3 | NP_004106.1 | |||
NM_175929.2 | 1835 | UTR 3 | NP_787125.1 |
ITGBL1 - integrin subunit beta like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |