Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATGTGGGCTGGCTAATTCTTGT[G/A]GGTGGTCCGTCCCGTGCACTGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615259 MIM: 190470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL21C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL21C - methyltransferase like 21C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010977.2 | Intron | NP_001010977.1 | ||||
XM_017020407.1 | Intron | XP_016875896.1 | ||||
XM_017020408.1 | Intron | XP_016875897.1 |
TPP2 - tripeptidyl peptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |