Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCTGTGGGTTTTTTTTCCCCCT[A/C]TGTTCCAATGTTACTAAAAACCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2AK4 - eukaryotic translation initiation factor 2 alpha kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013703.3 | Intron | NP_001013725.2 | ||||
XM_005254392.2 | Intron | XP_005254449.1 | ||||
XM_011521599.2 | Intron | XP_011519901.1 | ||||
XM_011521600.2 | Intron | XP_011519902.1 | ||||
XM_017022219.1 | Intron | XP_016877708.1 |