Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGTGCTTGGAGAAGGCTGGTG[A/G]GCTGTTCATGTTTTTTGAAGAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAHD1 - bromo adjacent homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301132.1 | Intron | NP_001288061.1 | ||||
NM_014952.4 | Intron | NP_055767.3 | ||||
XM_011521366.2 | Intron | XP_011519668.2 | ||||
XM_011521367.2 | Intron | XP_011519669.2 | ||||
XM_011521368.2 | Intron | XP_011519670.2 | ||||
XM_011521369.2 | Intron | XP_011519671.1 |