Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATGAGCTACTCTTTGTTGTTCC[A/C]TTCTTACCTCAGCAATCTGGGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370943 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105370943 - uncharacterized LOC105370943 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS18 - VPS18, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020857.2 | Intron | NP_065908.1 | ||||
XM_011521843.2 | Intron | XP_011520145.1 | ||||
XM_011521844.2 | Intron | XP_011520146.1 | ||||
XM_017022447.1 | Intron | XP_016877936.1 |