Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCACAGCACCTGCGGAGACTTT[A/G]CAAGGGACGTGTCAGAGCCGAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610169 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INO80 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INO80 - INO80 complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017553.2 | 3163 | UTR 3 | NP_060023.1 | |||
XM_011521685.2 | 3163 | Intron | XP_011519987.1 | |||
XM_011521686.2 | 3163 | UTR 3 | XP_011519988.1 |