Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACTCCTCCTCATCAAGCATGCC[A/G]TCGCAGTCGCAGTCGGCCAGCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605892 MIM: 605916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EHD4 - EH domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139265.3 | 1551 | Silent Mutation | GAC,GAT | D,D 496 | NP_644670.1 | |
XM_017022100.1 | 1551 | Silent Mutation | GAC,GAT | D,D 408 | XP_016877589.1 |
LOC105370792 - uncharacterized LOC105370792 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTBN5 - spectrin beta, non-erythrocytic 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |