Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTGGTTTGGGGACTTTATTAC[C/T]GGGTGGCCTTGGGCAAGTAACAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUOX1 - dual oxidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHF - Src homology 2 domain containing F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301168.1 | Intron | NP_001288097.1 | ||||
NM_001301169.1 | Intron | NP_001288098.1 | ||||
NM_001301170.1 | Intron | NP_001288099.1 | ||||
NM_001301171.1 | Intron | NP_001288100.1 | ||||
NM_138356.2 | Intron | NP_612365.2 | ||||
XM_006720742.3 | Intron | XP_006720805.2 | ||||
XM_011522174.2 | Intron | XP_011520476.1 | ||||
XM_011522175.2 | Intron | XP_011520477.1 | ||||
XM_011522176.2 | Intron | XP_011520478.1 | ||||
XM_011522177.2 | Intron | XP_011520479.1 | ||||
XM_011522178.2 | Intron | XP_011520480.1 | ||||
XM_017022710.1 | Intron | XP_016878199.1 | ||||
XM_017022711.1 | Intron | XP_016878200.1 |