Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTATCTTCCAGGTAGAAAGATGA[C/T]ATACTGGCACATTTCAATTTTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376714 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376714 - uncharacterized LOC105376714 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A4 - solute carrier family 30 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321036.1 | 6506 | Intron | NP_001307965.1 | |||
NM_013309.5 | 6506 | UTR 3 | NP_037441.2 | |||
XM_011521997.2 | 6506 | UTR 3 | XP_011520299.1 | |||
XM_017022560.1 | 6506 | UTR 3 | XP_016878049.1 | |||
XM_017022561.1 | 6506 | Intron | XP_016878050.1 | |||
XM_017022562.1 | 6506 | Intron | XP_016878051.1 |