Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTGACCGAAGAACTCGGCTGTG[A/G]TGAGATTATTGATAGAGAGTAGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605894 | ||||||||||||||||||||
Literature Links: |
EID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EID1 - EP300 interacting inhibitor of differentiation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014335.2 | 626 | Missense Mutation | GAT,GGT | D,G 181 | NP_055150.1 |
SHC4 - SHC adaptor protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203349.3 | 626 | Intron | NP_976224.3 | |||
XM_005254375.3 | 626 | Intron | XP_005254432.1 |