Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCAGAGGAAGGTTCCAGGCAT[C/G]ACAGTGCACATCGAGAAATATCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606910 MIM: 604447 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2L10 - BCL2 like 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNB5 - G protein subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006578.3 | 3173 | UTR 3 | NP_006569.1 | |||
NM_016194.3 | 3173 | UTR 3 | NP_057278.2 | |||
XM_011521162.2 | 3173 | UTR 3 | XP_011519464.1 | |||
XM_011521163.2 | 3173 | UTR 3 | XP_011519465.1 | |||
XM_017021867.1 | 3173 | UTR 3 | XP_016877356.1 |