Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATATTTAGACTTGCTTGCTATTT[A/G]GAAGATAATATGTACTAAGGTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIGB - phosphatidylinositol glycan anchor biosynthesis class B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004855.4 | Intron | NP_004846.4 | ||||
XM_005254795.4 | Intron | XP_005254852.1 | ||||
XM_011522235.2 | Intron | XP_011520537.1 | ||||
XM_011522236.2 | Intron | XP_011520538.1 | ||||
XM_011522237.2 | Intron | XP_011520539.1 | ||||
XM_017022730.1 | Intron | XP_016878219.1 | ||||
XM_017022731.1 | Intron | XP_016878220.1 | ||||
XM_017022732.1 | Intron | XP_016878221.1 |