Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACAAGGCCTGAGGGGACCCAT[A/G]TAGCTCCCCAGGGGCCCTGAGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCOM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCOM1 - GRINL1A complex locus 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018090.6 | Intron | NP_001018100.1 | ||||
NM_001018091.6 | Intron | NP_001018101.1 | ||||
NM_001285900.3 | Intron | NP_001272829.1 |
MYZAP - myocardial zonula adherens protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018100.4 | Intron | NP_001018110.1 | ||||
NM_152451.7 | Intron | NP_689664.3 |