Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGAATAGCTAAAAAATTTCAA[C/T]GTGAAGCAATTATCTAGGATTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ICE2 - interactor of little elongation complex ELL subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018089.2 | 4664 | UTR 3 | NP_001018099.1 | |||
NM_001276385.1 | 4664 | Intron | NP_001263314.1 | |||
NM_024611.5 | 4664 | UTR 3 | NP_078887.2 | |||
XM_005254661.1 | 4664 | UTR 3 | XP_005254718.1 | |||
XM_005254662.1 | 4664 | UTR 3 | XP_005254719.1 | |||
XM_011522009.1 | 4664 | UTR 3 | XP_011520311.1 | |||
XM_011522010.1 | 4664 | UTR 3 | XP_011520312.1 | |||
XM_011522011.2 | 4664 | Intron | XP_011520313.1 | |||
XM_017022569.1 | 4664 | UTR 3 | XP_016878058.1 |