Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTAGTACATAATAGATACAAAA[C/T]AAACAATAAAATGATATTTGTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGC15885 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGC15885 - uncharacterized protein MGC15885 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLN2 - talin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015059.2 | Intron | NP_055874.2 | ||||
XM_005254708.4 | Intron | XP_005254765.1 | ||||
XM_005254710.4 | Intron | XP_005254767.1 | ||||
XM_005254711.4 | Intron | XP_005254768.1 | ||||
XM_005254712.4 | Intron | XP_005254769.1 | ||||
XM_005254713.4 | Intron | XP_005254770.1 | ||||
XM_005254714.3 | Intron | XP_005254771.1 | ||||
XM_005254715.2 | Intron | XP_005254772.1 | ||||
XM_006720717.3 | Intron | XP_006720780.1 | ||||
XM_017022665.1 | Intron | XP_016878154.1 | ||||
XM_017022666.1 | Intron | XP_016878155.1 | ||||
XM_017022667.1 | Intron | XP_016878156.1 | ||||
XM_017022668.1 | Intron | XP_016878157.1 | ||||
XM_017022669.1 | Intron | XP_016878158.1 |