Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGATGTGCTGTCAGTTTTCCTTC[C/T]GGATGTACCAGAAACTGAGAGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612085 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASL12 - RAS like family 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307930.1 | 658 | Intron | NP_001294859.1 | |||
NM_016563.3 | 658 | Intron | NP_057647.1 | |||
XM_005254434.4 | 658 | Intron | XP_005254491.1 | |||
XM_011521660.2 | 658 | Intron | XP_011519962.1 | |||
XM_017022296.1 | 658 | Intron | XP_016877785.1 |
SLC51B - solute carrier family 51 beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178859.3 | 658 | Missense Mutation | CCG,CTG | P,L 121 | NP_849190.2 | |
XM_005254159.4 | 658 | Missense Mutation | CCG,CTG | P,L 121 | XP_005254216.1 |