Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGCGGGGTCACCTTGAGGCGG[G/T]TCAGCTGGTCGTGCAGGGCTGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176872 MIM: 180479 MIM: 605979 MIM: 609984 | ||||||||||||||||||||
Literature Links: |
MAP2K1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP2K1 - mitogen-activated protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4512 - microRNA 4512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL4 - ribosomal protein L4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNAPC5 - small nuclear RNA activating complex polypeptide 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD16 - small nucleolar RNA, C/D box 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD18A - small nucleolar RNA, C/D box 18A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD18B - small nucleolar RNA, C/D box 18B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD18C - small nucleolar RNA, C/D box 18C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZWILCH - zwilch kinetochore protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |