Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAGAGCACCCTGCAATGAGAAA[A/G]ATGCTGGATTTGGAGTCAGCCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612516 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UACA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UACA - uveal autoantigen with coiled-coil domains and ankyrin repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008224.1 | Intron | NP_001008225.1 | ||||
NM_018003.2 | Intron | NP_060473.2 | ||||
XM_005254529.4 | Intron | XP_005254586.1 | ||||
XM_011521752.2 | Intron | XP_011520054.2 | ||||
XM_011521753.2 | Intron | XP_011520055.2 | ||||
XM_011521754.2 | Intron | XP_011520056.1 | ||||
XM_017022394.1 | Intron | XP_016877883.1 | ||||
XM_017022395.1 | Intron | XP_016877884.1 |