Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACACAGGCTTTCGGGAGTGCAAG[A/G]GAAGTTCTTTAGCCCCACAAATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNUPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNUPN - snurportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042581.1 | 851 | Missense Mutation | CCT,TCT | P,S 236 | NP_001036046.1 | |
NM_001042588.1 | 851 | Missense Mutation | CCT,TCT | P,S 236 | NP_001036053.1 | |
NM_005701.3 | 851 | Missense Mutation | CCT,TCT | P,S 236 | NP_005692.1 |