Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCCTCAATTAATTTCATCATGT[A/G]ACTTGCTTGTTCAAAAATTTTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00597 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00597 - long intergenic non-protein coding RNA 597 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | Intron | NP_079052.2 | ||||
XM_005254670.2 | Intron | XP_005254727.1 | ||||
XM_005254671.2 | Intron | XP_005254728.1 | ||||
XM_005254673.2 | Intron | XP_005254730.1 | ||||
XM_005254674.2 | Intron | XP_005254731.1 | ||||
XM_005254675.4 | Intron | XP_005254732.1 | ||||
XM_011522033.2 | Intron | XP_011520335.1 | ||||
XM_011522034.2 | Intron | XP_011520336.1 | ||||
XM_011522035.2 | Intron | XP_011520337.1 | ||||
XM_011522036.2 | Intron | XP_011520338.1 | ||||
XM_011522037.2 | Intron | XP_011520339.1 | ||||
XM_011522038.2 | Intron | XP_011520340.1 | ||||
XM_011522039.2 | Intron | XP_011520341.1 | ||||
XM_011522040.1 | Intron | XP_011520342.1 | ||||
XM_011522041.1 | Intron | XP_011520343.1 | ||||
XM_011522042.2 | Intron | XP_011520344.1 |