Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTCTATGCACCTTTGCAAGAC[A/G]ACATAGCTTGTGTGACTGATGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605534 MIM: 614248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMG20A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMG20A - high mobility group 20A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304504.1 | Intron | NP_001291433.1 | ||||
NM_001304505.1 | Intron | NP_001291434.1 | ||||
NM_018200.3 | Intron | NP_060670.1 | ||||
XM_011521158.2 | Intron | XP_011519460.1 |
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | Intron | NP_079052.2 |