Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATTTCCCTGAATACAGAATA[C/T]GGCACAGAGAGATGGAGATTAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFL1 - elongation factor like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040610.2 | Intron | NP_001035700.1 | ||||
NM_001322844.1 | Intron | NP_001309773.1 | ||||
NM_001322845.1 | Intron | NP_001309774.1 | ||||
NM_024580.5 | Intron | NP_078856.4 | ||||
XM_011522000.1 | Intron | XP_011520302.1 |