Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCCCACCAGAAGCGTGAAGGAG[A/G]TGGTGATGTCAACAAGAACATATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC12A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC12A6 - solute carrier family 12 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042494.1 | 1567 | Missense Mutation | ACC,ATC | T,I 424 | NP_001035959.1 | |
NM_001042495.1 | 1567 | Missense Mutation | ACC,ATC | T,I 424 | NP_001035960.1 | |
NM_001042496.1 | 1567 | Missense Mutation | ACC,ATC | T,I 474 | NP_001035961.1 | |
NM_001042497.1 | 1567 | Missense Mutation | ACC,ATC | T,I 468 | NP_001035962.1 | |
NM_005135.2 | 1567 | Missense Mutation | ACC,ATC | T,I 432 | NP_005126.1 | |
NM_133647.1 | 1567 | Missense Mutation | ACC,ATC | T,I 483 | NP_598408.1 | |
XM_006720793.3 | 1567 | Missense Mutation | ACC,ATC | T,I 434 | XP_006720856.1 | |
XM_011522269.2 | 1567 | Missense Mutation | ACC,ATC | T,I 483 | XP_011520571.1 |