Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTATTTTTAATACACCATTTTCA[C/T]GCAGCCTAGTGACTGAAAACAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPH6 - diphthamine biosynthesis 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141972.1 | 1861 | Intron | NP_001135444.1 | |||
NM_080650.3 | 1861 | Intron | NP_542381.1 | |||
XM_011522160.1 | 1861 | Intron | XP_011520462.1 | |||
XM_017022707.1 | 1861 | Intron | XP_016878196.1 | |||
XM_017022708.1 | 1861 | UTR 3 | XP_016878197.1 | |||
XM_017022709.1 | 1861 | Intron | XP_016878198.1 |