Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAACCTGAACCTGACCCCTCATT[C/T]CTATACAGCAGTGGCTGTCACCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACAN - aggrecan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135.3 | Intron | NP_001126.3 | ||||
NM_013227.3 | Intron | NP_037359.3 | ||||
XM_006720419.1 | Intron | XP_006720482.1 | ||||
XM_011521313.1 | Intron | XP_011519615.1 | ||||
XM_011521314.1 | Intron | XP_011519616.1 | ||||
XM_017021985.1 | Intron | XP_016877474.1 | ||||
XM_017021986.1 | Intron | XP_016877475.1 | ||||
XM_017021987.1 | Intron | XP_016877476.1 |