Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAATGTTCTCTAAATCCCAACAC[A/G]TTCCATGCTCCCGGCTCTTAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603484 MIM: 608552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRC1 - protein regulator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS33B - VPS33B, late endosome and lysosome associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289148.1 | 2270 | UTR 3 | NP_001276077.1 | |||
NM_001289149.1 | 2270 | UTR 3 | NP_001276078.1 | |||
NM_018668.4 | 2270 | UTR 3 | NP_061138.3 | |||
XM_005254887.1 | 2270 | UTR 3 | XP_005254944.1 | |||
XM_011521448.1 | 2270 | UTR 3 | XP_011519750.1 | |||
XM_011521449.2 | 2270 | UTR 3 | XP_011519751.1 | |||
XM_017022075.1 | 2270 | UTR 3 | XP_016877564.1 | |||
XM_017022076.1 | 2270 | UTR 3 | XP_016877565.1 |