Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTACTGGCCAAAAGGTTCCAGC[A/T]GTCTGCTAAGGTGCCAAGCAGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGMA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGMA - repulsive guidance molecule family member a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166283.1 | 2162 | UTR 3 | NP_001159755.1 | |||
NM_001166286.1 | 2162 | UTR 3 | NP_001159758.1 | |||
NM_001166287.1 | 2162 | UTR 3 | NP_001159759.1 | |||
NM_001166288.1 | 2162 | UTR 3 | NP_001159760.1 | |||
NM_001166289.1 | 2162 | UTR 3 | NP_001159761.1 | |||
NM_020211.2 | 2162 | UTR 3 | NP_064596.2 |