Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGTCCCCGCGGGAAGCCGCGT[T/C]CCCCAGTCCCTTCCTGTGCGGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614147 MIM: 182455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF8 - C1q and tumor necrosis factor related protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207419.3 | 3283 | UTR 3 | NP_997302.2 | |||
XM_011522483.2 | 3283 | Intron | XP_011520785.1 |
SSTR5 - somatostatin receptor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSTR5-AS1 - SSTR5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |