Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCCAATGCTCACACAACCCCAG[A/C]AACTCTGGTCTGTTATTCCCATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G10 - phospholipase A2 group X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003561.2 | Intron | NP_003552.1 | ||||
XM_011522695.2 | Intron | XP_011520997.1 | ||||
XM_011522696.2 | Intron | XP_011520998.1 | ||||
XM_011522697.1 | Intron | XP_011520999.1 | ||||
XM_011522698.1 | Intron | XP_011521000.1 |