Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAGCCTCCACTGCGCATGTGCC[A/G]TGCATCCCTGCCCCTTCTGGGTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf45 - chromosome 16 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142469.1 | 1 | Intron | NP_001135941.1 | |||
NM_033201.2 | 1 | UTR 5 | NP_149978.1 | |||
XM_011522731.2 | 1 | Intron | XP_011521033.1 | |||
XM_017023824.1 | 1 | Intron | XP_016879313.1 |