Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATATGCACAGAACATGACACTAC[A/G]TAAAAACACAATGGAAAAAAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142240 MIM: 607782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HBQ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HBQ1 - hemoglobin subunit theta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LUC7L - LUC7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320226.1 | 1466 | Intron | NP_001307155.1 | |||
NM_018032.4 | 1466 | UTR 3 | NP_060502.1 | |||
NM_201412.2 | 1466 | Intron | NP_958815.1 | |||
XM_005255427.3 | 1466 | Intron | XP_005255484.1 | |||
XM_005255429.3 | 1466 | Intron | XP_005255486.1 | |||
XM_011522561.2 | 1466 | Intron | XP_011520863.1 | |||
XM_017023437.1 | 1466 | Intron | XP_016878926.1 | |||
XM_017023438.1 | 1466 | Intron | XP_016878927.1 | |||
XM_017023439.1 | 1466 | Intron | XP_016878928.1 | |||
XM_017023440.1 | 1466 | Intron | XP_016878929.1 |