Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACACTCCCTGTCAGCCTCCGACA[C/T]GGGCCTGCCCCAGAGGTCAGGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605213 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDPK1 - 3-phosphoinositide dependent protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261816.1 | 916 | Missense Mutation | ACG,ATG | T,M 402 | NP_001248745.1 | |
NM_002613.4 | 916 | Missense Mutation | ACG,ATG | T,M 402 | NP_002604.1 | |
NM_031268.5 | 916 | Missense Mutation | ACG,ATG | T,M 275 | NP_112558.2 | |
XM_011522521.2 | 916 | Missense Mutation | ACG,ATG | T,M 436 | XP_011520823.1 | |
XM_011522523.2 | 916 | Missense Mutation | ACG,ATG | T,M 309 | XP_011520825.1 | |
XM_017023271.1 | 916 | Missense Mutation | ACG,ATG | T,M 436 | XP_016878760.1 | |
XM_017023272.1 | 916 | Missense Mutation | ACG,ATG | T,M 404 | XP_016878761.1 | |
XM_017023273.1 | 916 | Missense Mutation | ACG,ATG | T,M 402 | XP_016878762.1 | |
XM_017023274.1 | 916 | Missense Mutation | ACG,ATG | T,M 375 | XP_016878763.1 | |
XM_017023275.1 | 916 | Missense Mutation | ACG,ATG | T,M 309 | XP_016878764.1 | |
XM_017023276.1 | 916 | Missense Mutation | ACG,ATG | T,M 275 | XP_016878765.1 | |
XM_017023277.1 | 916 | Missense Mutation | ACG,ATG | T,M 186 | XP_016878766.1 |