Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCATGAAGCACGGGGAACGGGT[C/T]GGATGGTAAGAGGCCAACCCAGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL21R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL21R - interleukin 21 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021798.3 | 629 | Intron | NP_068570.1 | |||
NM_181078.2 | 629 | Intron | NP_851564.1 | |||
NM_181079.4 | 629 | Silent Mutation | GTC,GTT | V,V 15 | NP_851565.4 | |
XM_011545857.2 | 629 | Silent Mutation | GTC,GTT | V,V 15 | XP_011544159.1 | |
XM_011545858.2 | 629 | UTR 5 | XP_011544160.1 | |||
XM_017023257.1 | 629 | Intron | XP_016878746.1 |