Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAATGAAAGACTCCGTGCTGC[C/G]TTTGGCATCAGTGATTCTTACGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRRM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRRM2 - serine/arginine repetitive matrix 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016333.3 | 941 | Silent Mutation | GCC,GCG | A,A 138 | NP_057417.3 | |
XM_005255226.1 | 941 | Silent Mutation | GCC,GCG | A,A 138 | XP_005255283.1 | |
XM_006720874.3 | 941 | Silent Mutation | GCC,GCG | A,A 138 | XP_006720937.1 |
SRRM2-AS1 - SRRM2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |